Skip to Content
Merck
All Photos(1)

Key Documents

380776

Sigma-Aldrich

1,3-Dichloro-2-(chloromethyl)-2-methylpropane

98%

Synonym(s):

1,1,1-Tris(chloromethyl)ethane

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€191.00

€191.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
€191.00

About This Item

Linear Formula:
CH3C(CH2Cl)3
CAS Number:
Molecular Weight:
175.48
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€191.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.482 (lit.)

bp

89-90 °C/22 mmHg (lit.)

mp

18 °C (lit.)

density

1.271 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

CC(CCl)(CCl)CCl

InChI

1S/C5H9Cl3/c1-5(2-6,3-7)4-8/h2-4H2,1H3

InChI key

BYXOMFFBGDPXHB-UHFFFAOYSA-N

General description

1,3-Dichloro-2-(chloromethyl)-2-methylpropane is a halogenated hydrocarbon. It is formed during the light induced reaction of diazomethane with chloroform.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photochemical Reactions of Diazomethane with Polyhalomethanes and a-Haloesters1.
Urry WH and Eiszner JR.
Journal of the American Chemical Society, 74(23), 5822-5826 (1952)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service