Skip to Content
Merck
All Photos(1)

Key Documents

380415

Sigma-Aldrich

Trimethyl(phenoxy)silane

97%

Synonym(s):

Phenoxy(trimethyl)silane, Phenyl trimethylsilyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OSi(CH3)3
CAS Number:
Molecular Weight:
166.29
Beilstein:
1905943
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

density

0.92 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)Oc1ccccc1

InChI

1S/C9H14OSi/c1-11(2,3)10-9-7-5-4-6-8-9/h4-8H,1-3H3

InChI key

OJAJJFGMKAZGRZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trimethyl(phenoxy)silane is a trimethylsilyl protected alcohol.[1] Direct lateral zincation (DlZn) of trimethyl(phenoxy)silane has been reported.[2] Dielectric studies of phenoxysilane-phenol binary systems has been reported.[3]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eva Hevia et al.
Chemical communications (Cambridge, England), (22)(22), 3240-3242 (2009-07-10)
A new application of zincate [(THF)Li(TMP)(tBu)Zn(tBu)] (1) in alkali-metal mediated zincation (AMMZn) is reported by isolating and structurally defining the first intermediates of direct lateral zincation (DlZn) of trimethyl(phenoxy)silane (2) both of which adopt novel six-element {ZnCSiOLiN} ring structures.
STUDIES ON ALCOXYSILANE-ALCOHOL AND PHENOXYSILANE-PHENOL BINARY SYSTEMS.
Nagy J, et al.
Chemical Engineering Journal, 15(3), 155-167 (1971)
Trimethylsilyl Protection of Alcohols Over Phosphorus Pentoxide Supported on Silica Gel.
Shaterian HR, et al.
Phosphorus, Sulfur, and Silicon and the Related Elements, 186(7), 1604-1611 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service