Skip to Content
Merck
All Photos(2)

Key Documents

377392

Sigma-Aldrich

(R)-(−)-Citronellyl bromide

95%

Synonym(s):

8-Bromo-2,6-dimethyl-2-octene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2CH(CH3)CH2CH2Br
CAS Number:
Molecular Weight:
219.16
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

optical activity

[α]20/D −6.8°, neat

refractive index

n20/D 1.474 (lit.)

bp

111 °C/12 mmHg (lit.)

density

1.11 g/mL at 25 °C (lit.)

functional group

alkyl halide
bromo

SMILES string

C[C@@H](CCBr)CC\C=C(/C)C

InChI

1S/C10H19Br/c1-9(2)5-4-6-10(3)7-8-11/h5,10H,4,6-8H2,1-3H3/t10-/m1/s1

InChI key

QPKCDMXLSDFCQD-SNVBAGLBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(R)-(−)-Citronellylbromide can be used as a reactant to synthesize:
  • (R)-3,7-dimethyloct-6-ene-1-amine via Gabriel synthesis which is then condensed with dicarboxylic acids to synthesize retro amides.
  • Citronellyl derivatives of sulfoximines and sulfondiimines via alkylation using potassium hydroxide and dimethyl sulfoxide.
  • Citronellyltriphenylphosphonium bromide which can be used as a key intermediate to synthesize (+)-caparratriene via Witting reaction.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective Synthesis of the Antileukemic Sesquiterpene (+)-Caparratriene from L-menthol and Tiglic Aldehyde
Vydrina V, et al.
Chemistry of Natural Compounds, 54, 461-463 (2018)
N-Alkylations of NH-Sulfoximines and NH-Sulfondiimines with Alkyl Halides Mediated by Potassium Hydroxide in Dimethyl Sulfoxide
Hendriks C, et al.
advanced synthesis and catalysis, 356, 1847-1852 (2014)
Takuho Saito et al.
Organic & biomolecular chemistry, 18(21), 3996-3999 (2020-05-16)
Inversion of the connectivity of amide groups in foldable azobenzene dyads with chiral side chains, which can self-assemble into toroids and nanotubes, significantly increases the thermal stability of these aggregates. The results can be explained by the geometrical difference of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service