377090
(−)-2,3-O-Isopropylidene-D-erythronolactone
98%
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Assay
98%
optical activity
[α]20/D −118°, c = 1 in H2O
mp
67-69 °C (lit.)
SMILES string
CC1(C)O[C@@H]2COC(=O)[C@@H]2O1
InChI
1S/C7H10O4/c1-7(2)10-4-3-9-6(8)5(4)11-7/h4-5H,3H2,1-2H3/t4-,5-/m1/s1
InChI key
WHPSMBYLYRPVGU-RFZPGFLSSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Undergoes Aldol condensations with silyl ketene acetals. Employed in spiroannulated carbohydrate synthesis. Convergent syntheses of a hydroxylated indolizidine, carbohydrate substituted benzoquinones, and of the oxazole segment of calyculin have been accomplished using this chiral synthon.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Journal of Organic Chemistry, 54, 2032-2032 (1989)
Tetrahedron, 50, 3333-3333 (1994)
Tetrahedron, 50, 1111-1111 (1994)
The Journal of Organic Chemistry, 54, 2591-2591 (1989)
Tetrahedron Letters, 32, 4859-4859 (1991)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service