Skip to Content
Merck
All Photos(1)

Documents

376825

Sigma-Aldrich

2-Chloro-5-(trifluoromethyl)benzaldehyde

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CF3)CHO
CAS Number:
Molecular Weight:
208.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.488 (lit.)

bp

42-44 °C/1.5 mmHg (lit.)

density

1.435 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)c1ccc(Cl)c(C=O)c1

InChI

1S/C8H4ClF3O/c9-7-2-1-6(8(10,11)12)3-5(7)4-13/h1-4H

InChI key

OZZOJJJYKYKBNH-UHFFFAOYSA-N

Application

2-Chloro-5-(trifluoromethyl)benzaldehyde may be used as reactant in the three-component reaction of 2-halogenated aromatic aldehydes, 1H-benzo[d]imidazol-5-amine and cyclohexane-1,3-diones to afford imidazoquinolinoacridinone derivatives. It may be used in the gas-phase chemical derivatization of poly(tetrafluoroethylene) and poly(ethylene terephthalate) using gas plasma.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Domino synthesis of fused hexacyclic imidazoquinolinoacridinones catalyzed by CuI/l-proline.
Li C, et al.
Tetrahedron, 70(46), 8919-8924 (2014)
Surface chemical derivatization of plasma-treated PET and PTFE.
Markkula TK, et al.
Surface and Interface Analysis : SIA, 34(1), 583-587 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service