Skip to Content
Merck
All Photos(2)

Key Documents

375691

Sigma-Aldrich

2-Hydroxyimino-2-phenylacetonitrile, mixture of syn and anti

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€53.60

€53.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€53.60

About This Item

Linear Formula:
C6H5C(=NOH)CN
CAS Number:
Molecular Weight:
146.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€53.60


Check Cart for Availability

Request a Bulk Order

Assay

97%

mp

128-130 °C (lit.)

functional group

amine
nitrile
oxime
phenyl

SMILES string

ON=C(C#N)c1ccccc1

InChI

1S/C8H6N2O/c9-6-8(10-11)7-4-2-1-3-5-7/h1-5,11H

InChI key

MJCQFBKIFDVTTR-UHFFFAOYSA-N

Related Categories

Application

2-Hydroxyimino-2-phenylacetonitrile may be used in the synthesis of 2-t-butoxycarbonyloxyimino-2-phenylacetonitrile[1] and o-chlorobenzyloxycarbonylimino-2-phenylacetonitrile.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Scott Bohle et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(13), 4223-4229 (2013-02-13)
The reaction of nitric oxide with benzyl cyanide in the presence of potassium methoxide at low temperature gave the dipotassium salt of a bis-diazeniumdiolate 2 in excellent yield. Two new stereospecific syntheses of E or Z 2-(hydroxyimino)-2-phenylacetonitrile from 2 have
Refinement of the crystal structure of 2-hydroxyimino-2-phenylacetonitrile, C8H6N2O, at 173 ?.
Razzino P, et al.
Zeitschrift fur Kristallographie - New Crystal Structures, 216(1-4), 583-584 (2001)
A facile method for the preparation of oxime t-butyl carbonates for evaluation of the Beckman rearrangement.
Mamaghani M and Samimi HA.
Orient. J. Chem., 24(3), 837-841 (2008)
THE SYNTHESIS OF j> METHOXYBENZYLOXYCARBONYL AMINO ACIDS.
Wang K-T.
J. Chin. Chem. Soc., 34, 117-123 (1987)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service