Skip to Content
Merck
All Photos(1)

Key Documents

375209

Sigma-Aldrich

Trimethyltin chloride solution

1.0 M in methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SnCl
CAS Number:
Molecular Weight:
199.27
Beilstein:
3535111
MDL number:
UNSPSC Code:
12352103

concentration

1.0 M in methylene chloride

density

1.356 g/mL at 25 °C

SMILES string

C[Sn](C)(C)Cl

InChI

1S/3CH3.ClH.Sn/h3*1H3;1H;/q;;;;+1/p-1

InChI key

KWTSZCJMWHGPOS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged under nitrogen in Sure/Seal bottles.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

comparable product


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flaviana T Vieira et al.
European journal of medicinal chemistry, 45(3), 883-889 (2009-12-05)
The reaction of 1,3-cyclohexadione with 2-aminobenzoic acid has produced the 2-(3-oxocyclohex-1-enyl)benzoic acid (HOBz). Subsequent reactions of the ligand with organotin chlorides led to [Me(2)Sn(OBz)O](2) (1), [Bu(2)Sn(OBz)O](2) (2), [Ph(2)Sn(OBz)O](2) (3), [Me(3)Sn(OBz)] (4), [Bu(3)Sn(OBz)] (5) and [Ph(3)Sn(OBz)] (6). All complexes have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service