Skip to Content
Merck
All Photos(2)

Key Documents

375098

Sigma-Aldrich

6-Chloroquinoline

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6ClN
CAS Number:
Molecular Weight:
163.60
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

126-127 °C/10 mmHg (lit.)

mp

41-43 °C (lit.)

functional group

chloro

SMILES string

Clc1ccc2ncccc2c1

InChI

1S/C9H6ClN/c10-8-3-4-9-7(6-8)2-1-5-11-9/h1-6H

InChI key

GKJSZXGYFJBYRQ-UHFFFAOYSA-N

General description

6-Chloroquinoline is a haloquinoline. Pd/C-catalyzed Suzuki-Miyaura coupling of 6-chloroquinoline with 2-(dicyclohexylphosphino)biphenyl is reported.[1] The polarographic behavior of 6-chloroquinoline in DMF solutions and mechanism of its reduction at the dropping mercury electrode has been reported.[2] Stability constants of molecular iodine complexes of 6-chloroquinoline in aliphatic and aromatic hydrocarbons, carbon tetrachloride, bromobenzene and chloro-substituted benzenes was investigated by spectrophotometric methods.[3]
The standard molar enthalpy of formation of 6-chloroquinoline has been derived from the standard molar enthalpy of combustion and was evaluated in terms of molecular structure.[4] Its arylation reaction with diamine derivatives of adamantanes in the presence of palladium catalyst has been reported to afford N,N′-diaryl derivatives.[5]

Application

6-Chloroquinoline may be used in the synthesis of 3-methyl-3H-imidazo[4,5-f]quinolin-2-amine.[6] It may be used as catalyst for high yield preparation of ethoxycarbonyl isothiocyanate.[7]
Catalyst for high yield preparation of ethoxycarbonyl isothiocyanate.[7]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Standard molar enthalpies of formation of 2-chloroquinoline, 4-chloroquinoline, 6-chloroquinoline and 4, 7-dichloroquinoline by rotating-bomb calorimetry.
da Silva MAVR, et al.
The Journal of Chemical Thermodynamics, 38(1), 49-55 (2006)
The Journal of Organic Chemistry, 55, 5230-5230 (1990)
Arylation of adamantanamines: V. Palladium-catalyzed amination of isomeric chloroquinolines with diamines of the adamantane series.
Grigorova OK, et al.
Russ. J. Org. Chem., 48(12), 1495-1508 (2012)
Tsuyoshi Tagata et al.
The Journal of organic chemistry, 68(24), 9412-9415 (2003-11-25)
A phosphine ligand, such as PPh3 or 2-(dicyclohexylphosphino)biphenyl, is essential for the Pd/C-catalyzed Suzuki-Miyaura coupling of halopyridines and haloquinolines, although it has been reported that the reaction of phenyl chlorides can be catalyzed by nonprereduced Pd/C without any additives. In
Solvent effect on thermodynamic stability of iodine complexes with quinoline and pyridine bases.
Uruska I.
Spectrochimica Acta Part A: Molecular Spectroscopy, 36(7), 639-646 (1980)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service