Skip to Content
Merck
All Photos(2)

Key Documents

375047

Sigma-Aldrich

4-(Chloromethyl)phenyl isocyanate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€189.00

€189.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€189.00

About This Item

Linear Formula:
ClCH2C6H4NCO
CAS Number:
Molecular Weight:
167.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€189.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

32-33 °C (lit.)

density

1.216 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ClCc1ccc(cc1)N=C=O

InChI

1S/C8H6ClNO/c9-5-7-1-3-8(4-2-7)10-6-11/h1-4H,5H2

InChI key

OBYMZHLTKVXPQF-UHFFFAOYSA-N

General description

4-(Chloromethyl)phenyl isocyanate is an aryl isocyanate. It is reported to participate in the anomeric O-acylation of Kdo.[1]

Application

4-(Chloromethyl)phenyl isocyanate may be used in the synthesis of series of carbamate derivatives of Hoechst 33258, potential anticancer agents.[2]

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsuyoshi Ichiyanagi et al.
Carbohydrate research, 340(17), 2682-2687 (2005-10-04)
To develop a convenient method for the preparation of an alpha-Kdo derivative carrying a functional spacer at the reducing end, we examined anomeric O-acylation using Kdo and halogenated alkyl/aryl isocyanates as nucleophile and electrophiles, respectively. Reaction of a Kdo derivative
Krzysztof Bielawski et al.
Biological & pharmaceutical bulletin, 25(7), 916-919 (2002-07-23)
A series of carbamate derivatives of Hoechst 33258 was prepared as potential anticancer agents. These new compounds (1-4) were readily prepared in good yields by addition of chloroethyl, bromoethyl, chloropropyl or 4-(chloromethyl)phenyl isocyanates to Hoechst 33258. Their cytotoxic activity was
Ashraf Ali et al.
Journal of separation science, 40(12), 2654-2661 (2017-05-06)
A specially designed long open tubular capillary column (50 μm internal diameter and 112 cm effective length) was prepared by fabrication of a thin three-component co-polymer layer on the inner surface of silica capillary. A pretreated silica capillary was reacted with 4-(chloromethyl)phenyl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service