Skip to Content
Merck
All Photos(2)

Documents

374172

Sigma-Aldrich

Ethyl ethoxyacetate

98%

Synonym(s):

Diethyl glycolate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OCH2COOCH2CH3
CAS Number:
Molecular Weight:
132.16
Beilstein:
1749545
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.402 (lit.)

bp

156 °C (lit.)

density

0.975 g/mL at 25 °C (lit.)

SMILES string

CCOCC(=O)OCC

InChI

1S/C6H12O3/c1-3-8-5-6(7)9-4-2/h3-5H2,1-2H3

InChI key

CKSRFHWWBKRUKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl ethoxyacetate is an ester. It is reported to participate in the ozonation of ethers.

Application

Ethyl ethoxyacetate may be used in the preparation of ethoxyacetylacetone.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pyridine Derivatives. I. 3-Cyano-4-ethoxymethyl-6-methyl-2-pyridone and some related transformation products.
Bruce WF and Coover Jr HW.
Journal of the American Chemical Society, 66(12), 2092-2094 (1944)
The course of ozonation of ethers.
Price CC and Tumolo AL.
Journal of the American Chemical Society, 86(21), 4691-4694 (1964)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service