Skip to Content
Merck
All Photos(1)

Key Documents

373710

Sigma-Aldrich

Molybdenum(VI) dichloride dioxide

Synonym(s):

Dichlorodioxomolybdenum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MoO2Cl2
CAS Number:
Molecular Weight:
198.84
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
flakes
Pricing and availability is not currently available.

grade

for analytical purposes

Quality Level

form

flakes

concentration

34.4-36.9% chloride (silver nitrate titration)

density

3.31 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

Cl[Mo](Cl)(=O)=O

InChI

1S/2ClH.Mo.2O/h2*1H;;;/q;;+2;;/p-2

InChI key

JCFNAADCQWXIJD-UHFFFAOYSA-L

General description

Molybdenum(VI) dichloride dioxide is an oxo-halide of molybdenum widely used as a catalyst for various organic transformations. It is an oxo-transfer catalyst that has the ability to promote oxidation as well as reduction reactions. It can also be used as a chemical vapor deposition(CVD) precursor to synthesize various Mo-based nanomaterials.[1] [2]

Application

[3]Molybdenum(VI) dichloride dioxide can be used as a catalyst:
  • For thioacetalization of heterocyclic, aromatic, and aliphatic compounds.[1]
  • To synthesize mono-, di-, tri-, and tetracarbamates from alcohols and aromatic or aliphatic isocyanates.[4]
  • For stereospecific deoxygenation of epoxides to alkenes.[5]
  • For the synthesis of dihydropyrimidinone/thione and polyhydroquinoline derivatives via a Biginelli and Hantzsch reaction.[3]
It can also be used as a precursor to synthesize MoO2 nanoparticles.[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interplay between size and crystal structure of molybdenum dioxide nanoparticles?synthesis, growth mechanism, and electrochemical performance
Dorota Koziej, et al.
Small, 7, 377-387 (2011)
Mild and high-yielding molybdenum (VI) dichloride dioxide-catalyzed formation of mono-, di-, tri-, and tetracarbamates from alcohols and aromatic or aliphatic isocyanates
Christian Stock, et al.
advanced synthesis and catalysis, 354, 2309-2330 (2012)
Molybdenum pentachloride (MoCl5) or molybdenum dichloride dioxide (MoO2Cl2): advanced catalysts for thioacetalization of heterocyclic, aromatic and aliphatic compounds
Shyamaprosad Goswami and Annada C. Maity
Tetrahedron Letters, 49, 3092-3096 (2008)
Molybdenum-Catalyzed Stereospecific Deoxygenation of Epoxides to Alkenes
Sobi Asako, et al.
advanced synthesis and catalysis, 358, 3966-3970 (2016)
MoO2Cl2 catalyzed efficient synthesis of functionalized 3,4-dihydropyrimidin-2 (1 H)-ones/thiones and polyhydroquinolines: recyclability, fluorescence and biological studies
Sravanthi Devi Guggilapu, et al.
Journal of Chemistry, 40, 838-843 (2016)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service