Skip to Content
Merck
All Photos(1)

Key Documents

373567

Sigma-Aldrich

Ruthenium(III) nitrosyl nitrate solution

in dilute nitric acid

Synonym(s):

Nitrilooxonium ruthenium(2+) nitrate (1:1:3)

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€87.60
250 ML
€541.00

€87.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
€87.60
250 ML
€541.00

About This Item

Linear Formula:
Ru(NO)(NO3)x(OH)y, x+y=3
CAS Number:
Molecular Weight:
318.10
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
liquid

€87.60


Check Cart for Availability

Request a Bulk Order

grade

for analytical purposes

Quality Level

form

liquid

composition

Ru, 1.5% (typical)

reaction suitability

reagent type: catalyst
core: ruthenium

concentration

in dilute nitric acid

density

1.07 g/mL at 25 °C

SMILES string

[Ru+3]N=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/3NO3.NO.Ru/c3*2-1(3)4;1-2;/q4*-1;+4

InChI key

VAAILMLOAHTPQK-UHFFFAOYSA-N

General description

Ruthenium(III) nitrosyl nitrate solution is a stable and water-soluble compound. The presence of the nitrosyl ligand contributes to its distinct reactivity and coordination behavior. Additionally, the compound exhibits characteristic spectroscopic properties that make it valuable in analytical and research applications.

Application

Ruthenium(III) nitrosyl nitrate solution can be used:
  • As a Ru source to prepare Y2Ru2O7/NiMoO4@NF electrocatalyst by a combined sol–gel and hydrothermal method, which exhibits superior stability for overall water splitting in an alkaline medium.
  • To fabricate bifunctional electrode for Li-air batteries.
  • As a precursor to synthesize ruthenium nanoparticles with variable sizes. It helps to form stable ruthenium nitrite complex avoiding the unwanted metal oxide hydrolysis or precipitation.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Hydrogen is one of the most important resources in providing food, fuel, and chemical products for our everyday life. Sustainable catalytic hydrogen production from bioethanol has gained significant attention in recent years due to globally diminishing fossil fuel supplies, which have necessitated the search for new chemical feedstocks.

Questions

1–3 of 3 Questions  
  1. 구성: Ru1.5%라고 적혀있는데, 전체 시약에 대한 weight ratio인가요 아니면 molar ratio인가요?

    1 answer
    1. The composition "Ru 1.5%" refers to the weight ratio of ruthenium (Ru) in the entire reagent.

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service