Skip to Content
Merck
All Photos(1)

Key Documents

372099

Sigma-Aldrich

2,4-Di-tert-amylphenol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C2H5C(CH3)2]2C6H3OH
CAS Number:
Molecular Weight:
234.38
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

bp

169-170 °C/22 mmHg (lit.)

mp

25 °C (lit.)

density

0.930 g/mL at 25 °C (lit.)

SMILES string

CCC(C)(C)c1ccc(O)c(c1)C(C)(C)CC

InChI

1S/C16H26O/c1-7-15(3,4)12-9-10-14(17)13(11-12)16(5,6)8-2/h9-11,17H,7-8H2,1-6H3

InChI key

WMVJWKURWRGJCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The standard molar enthalpies of sublimation (or vaporization) of 2,4-di-tert-amylphenol have been evaluated. Preparation of 2,4-di-tert-amylphenol, via catalytic alkylation of phenol with trimethylethylene, is reported.

Application

2,4-Di-tert-amylphenol is a phenolic compound, used for the laccase induced coupling reactions. It may be used in the synthesis of Cu-phthalocyanine-C60.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular association and monolayer formation of soluble phthalocyanine compounds.
Snow AW and Jarvis NL.
Journal of the Chemical Society, 106(17), 4706-4711 (1984)
Thermochemistry of phenols: quantification of the< i> ortho-,< i> para</i>-, and< i> meta</i>-interactions in tert-alkyl substituted phenols.
Verevkin SP.
The Journal of Chemical Thermodynamics, 31(5), 559-585 (1999)
Molecular association and monolayer formation of soluble phthalocyanine compounds.
Viktorova EA, et al.
Russian Chemical Bulletin, 9(8), 1402-1402 (1960)
Enzymatic synthesis of Tinuvin.
Schroeder M, et al.
Enzyme and Microbial Technology, 40(7), 1748-1752 (2007)
Yan-Hua Liu et al.
Ecotoxicology and environmental safety, 135, 90-97 (2016-10-11)
The occurrence and distribution of eight selected endocrine-disrupting chemicals were investigated in samples of surface water and suspended particulate matter (SPM) in Nanjing section of Yangtze River over a year (the flow period, the wet period and the dry period).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service