Skip to Content
Merck
All Photos(1)

Key Documents

371491

Sigma-Aldrich

Thionyl chloride

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SOCl2
CAS Number:
Molecular Weight:
118.97
Beilstein:
1209273
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
technical grade
form:
liquid
Pricing and availability is not currently available.

grade

technical grade

vapor pressure

97 mmHg ( 20 °C)

form

liquid

refractive index

n20/D 1.518 (lit.)

bp

79 °C (lit.)

mp

−105 °C (lit.)

density

1.631 g/mL at 25 °C (lit.)

SMILES string

ClS(Cl)=O

InChI

1S/Cl2OS/c1-4(2)3

InChI key

FYSNRJHAOHDILO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Aksoy et al.
Journal of biotechnology, 60(1-2), 37-46 (1998-05-08)
Poly(methyl methacrylate-acrylic acid) microspheres were prepared and the acid groups were activated by using either carbodiimide (CDI) or thionyl chloride (SOCl2). alpha-Amylase was covalently bound on these activated microspheres. The properties of the immobilized enzyme were investigated and compared with
N L Benoiton et al.
International journal of peptide and protein research, 43(4), 321-324 (1994-04-01)
Fmoc-glutamic acid is converted by thionyl chloride into the dichloride, which spontaneously cyclizes to Fmoc-pyroglutamyl chloride. The latter is stable to water. Pure Fmoc-pyroglutamyl chloride is obtained by washing the reaction mixture with water, which destroys uncyclized dichloride by converting
Je Seob Park et al.
ACS applied materials & interfaces, 3(2), 360-368 (2011-01-07)
In this study, we present a facile means of fabricating graphene thin films via layer-by-layer (LbL) assembly of charged graphene nanosheets (GS) based on electrostatic interactions. To this end, graphite oxide (GO) obtained from graphite powder using Hummers method is
R Karunanithy et al.
Journal of pharmacobio-dynamics, 12(8), 468-475 (1989-08-01)
The relationships between contraceptive activity, hydrophobic character and alkaline hydrolysis rates were studied in three homologous series of norethisterone and levonorgestrel esters. Hydrophobic character was expressed by the high-performance liquid chromatographic retention term, VR(w) or retention volume at 100% aqueous
Xiangshu Xiao et al.
The Journal of organic chemistry, 70(16), 6496-6498 (2005-07-30)
The indeno[1,2-c]isoquinolines are an important class of topoisomerase I inhibitors with anticancer activity. The condensation of Schiff bases with homophthalic anhydrides provides a mixture of cis- and trans-4-carboxy-3,4-dihydro-3-phenyl-1(2H)isoquinolones. Although the cis products can be readily converted to indeno[1,2-c]isoquinolines with thionyl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service