Skip to Content
Merck
All Photos(1)

Key Documents

370150

Sigma-Aldrich

Methyl (2-chloro-1-methylethylidene)hydrazinecarboxylate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9ClN2O2
CAS Number:
Molecular Weight:
164.59
MDL number:

Assay

97%

SMILES string

COC(=O)N\N=C(/C)CCl

InChI

1S/C5H9ClN2O2/c1-4(3-6)7-8-5(9)10-2/h3H2,1-2H3,(H,8,9)/b7-4+

InChI key

LCGQSEGTIXIWOF-QPJJXVBHSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service