369780
Diethoxyphosphinyl isocyanate
96%
Synonym(s):
Diethyl phosphorisocyanatidate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C2H5O)2P(O)NCO
CAS Number:
Molecular Weight:
179.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
96%
form
liquid
refractive index
n20/D 1.416 (lit.)
bp
85-87 °C/10 mmHg (lit.)
density
1.165 g/mL at 25 °C (lit.)
SMILES string
CCOP(=O)(OCC)N=C=O
InChI
1S/C5H10NO4P/c1-3-9-11(8,6-5-7)10-4-2/h3-4H2,1-2H3
InChI key
BSCBJDQIQBYMBT-UHFFFAOYSA-N
General description
Diethoxyphosphinyl isocyanate is an isocyanate derivative. It has been investigated for its performance as film forming electrolyte additive in propylene carbonate (PC) and EC/EMC-based electrolytes.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
118.4 °F - closed cup
Flash Point(C)
48 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
M Nehéz et al.
Ecotoxicology and environmental safety, 17(2), 258-263 (1989-04-01)
Phenyl isocyanate (I) and diethoxyphosphoryl isocyanate (II), used as intermediates in organic chemical syntheses, were tested for their acute toxic, cytogenetic, and embryotoxic activity on mice of different strains. The oral LD50 values for male CFLP mice were determined to
G Brigden et al.
European journal of nuclear medicine, 16(11), 795-799 (1990-01-01)
A new, single bolus method of in vivo blood pool imaging using a technetium Tc99m phosphine isocyanide complex (DEPIC) which binds to pre-albumin was evaluated in volunteers (n = 4) and patients (n = 20). DEPIC was assessed for its
Isocyanate compounds as electrolyte additives for lithium-ion batteries.
Korepp C, et al.
Journal of Power Sources, 174(2), 387-393 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service