Skip to Content
Merck

Skip To

369705

Poly(4-tert-butylstyrene)

average Mw 50,000-100,000

Synonym(s):

PTBS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 g
Check Cart for Availability
€ 128,00

About This Item

Linear Formula:
[CH2CH[C6H4C(CH3)3]]n
CAS Number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
MDL number:

€ 128,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

mol wt

average Mw 50,000-100,000

SMILES string

CC(C)(C)c1ccc(C=C)cc1

InChI

1S/C12H16/c1-5-10-6-8-11(9-7-10)12(2,3)4/h5-9H,1H2,2-4H3

InChI key

QEDJMOONZLUIMC-UHFFFAOYSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
553468ALD0028808057
form

solid

form

solid

form

solid

form

tablet

mol wt

average Mw 50,000-100,000

mol wt

-

mol wt

-

mol wt

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters