Skip to Content
Merck
All Photos(1)

Key Documents

369640

Sigma-Aldrich

2,4-Dibromo-2,4-dimethyl-3-pentanone

99%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€105.00

€105.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€105.00

About This Item

Linear Formula:
BrC(CH3)2COC(CH3)2Br
CAS Number:
Molecular Weight:
271.98
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€105.00


Check Cart for Availability

Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.506 (lit.)

bp

89-91 °C/13 mmHg (lit.)

density

1.61 g/mL at 25 °C (lit.)

functional group

bromo
ketone

SMILES string

CC(C)(Br)C(=O)C(C)(C)Br

InChI

1S/C7H12Br2O/c1-6(2,8)5(10)7(3,4)9/h1-4H3

InChI key

SWSOAIMZWLDPST-UHFFFAOYSA-N

General description

2,4-Dibromo-2,4-dimethyl-3-pentanone is an α,α′-dibromoketone. Electroreduction of 2,4-dibromo-2,4-dimethyl-3-pentanone in polar aprotic solvent has been reported.[1] Electrochemical reduction of 2,4-dibromo-2,4-dimethyl-3-pentanone in acetic acid and sodium actetate yields α-acetoxy ketones.[2] 2,4-Dibromo-2,4-dimethyl-3-pentanone reacts with lithium dimethylcuprate(I) in diethyl ether at -78°C to yield 2,2,4-trimethyl-3-pentanone (an α methyl ketone).[3]

Application

2,4-Dibromo-2,4-dimethyl-3-pentanone may be used in the synthesis of 2-dimethylamino-4-methylene-1,3-dioxolanes, via debromination using zinc-copper couple in dimethylformamide and dimethylacetamide.[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Debromination of. alpha.,. alpha. 1-dibromo ketones with a zinc-copper couple in dimethylformamide and dimethylacetamide. New reaction yielding 2-dimethylamino-4-methylene-1, 3-dioxolanes.
Hoffmann HMR, et al.
Journal of the American Chemical Society, 94(9), 3201-3204 (1972)
Electrochemical reduction of. alpha.,. alpha.'-dibromo ketones in acetic acid. Convenient synthetic route to highly branched. alpha.-acetoxy ketones.
Fry AJ and O'Dea JJ.
The Journal of Organic Chemistry, 40(25), 3625-3631 (1975)
Reaction of. alpha.,. alpha.'-dibromo ketones with organocopper reagents. New method for. alpha. alkylation of ketones.
Posner GH, et al.
Journal of the American Chemical Society, 95(9), 3076-3077 (1973)
Electroreduction of. alpha.,. alpha.'-dibromoketones. 2, 4-Dibromo-2, 4-dimethyl-3-pentanone.
Dirlam JP, et al.
Journal of the American Chemical Society, 94(1), 240-245 (1972)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service