Skip to Content
Merck
All Photos(2)

Documents

36815

Sigma-Aldrich

1-(Diethoxymethyl)imidazole

≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14N2O2
CAS Number:
Molecular Weight:
170.21
Beilstein:
908482
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0%

form

liquid

refractive index

n20/D 1.464

density

1.041 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(OCC)n1ccnc1

InChI

1S/C8H14N2O2/c1-3-11-8(12-4-2)10-6-5-9-7-10/h5-8H,3-4H2,1-2H3

InChI key

NNQBAGFIYBBAFL-UHFFFAOYSA-N

General description

1-(Diethoxymethyl)imidazole is an orthoamide protected imidazole. The dianion reacts selectively at the 2 position 1-(Diethoxymethyl)imidazole is a 1-substituted imidazoles. Isoprene-mediated lithiation of 1-(diethoxymethyl)imidazole, leads to the preparation of the corresponding 2-lithio intermediate, which reacts with different electrophiles to afford 2-functionalised imidazoles.

Application

1-(Diethoxymethyl)imidazole may be used in the preparation of various hydroxy- and amino-functionalized imidazoles, via isoprene-mediated lithiation followed by reaction with an electrophile.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

233.6 °F

Flash Point(C)

112.0 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1, 2-Functionalized Imidazoles as Palladium Ligands: An Efficient and Robust Catalytic System for the Fluorine-Free Hiyama Reaction.
Martinez R, et al.
European Journal of Organic Chemistry, 4, 872-877 (2014)
N.J. Curtis et al.
The Journal of Organic Chemistry, 45, 4038-4038 (1980)
Isoprene-catalysed lithiation: deprotection and functionalisation of imidazole derivatives.
Torregrosa R, et al.
Tetrahedron, 63(4), 947-952 (2007)
L.N. Pridgen et al.
Tetrahedron Letters, 38, 1275-1275 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service