Skip to Content
Merck
All Photos(1)

Key Documents

367664

Sigma-Aldrich

1,1,1,3,3,3-Hexafluoroisopropyl methacrylate

99%

Synonym(s):

HFiPMA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CO2CH(CF3)2
CAS Number:
Molecular Weight:
236.11
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

0.71 psi ( 20 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.331 (lit.)

density

1.302 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(=O)OC(C(F)(F)F)C(F)(F)F

InChI

1S/C7H6F6O2/c1-3(2)4(14)15-5(6(8,9)10)7(11,12)13/h5H,1H2,2H3

InChI key

FMQPBWHSNCRVQJ-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service