36755
N-Methyldidecylamine
95%
Synonym(s):
DAMA-10, Didecylmethylamine
Select a Size
About This Item
Recommended Products
Quality Level
Assay
95%
form
liquid
impurities
≤0.5% water
refractive index
n20/D 1.448
density
0.807 g/mL at 20 °C (lit.)
functional group
amine
SMILES string
CCCCCCCCCCN(C)CCCCCCCCCC
InChI
1S/C21H45N/c1-4-6-8-10-12-14-16-18-20-22(3)21-19-17-15-13-11-9-7-5-2/h4-21H2,1-3H3
InChI key
ATBNMWWDBWBAHM-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
200.1 °F - closed cup
Flash Point(C)
93.4 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service