Skip to Content
Merck
All Photos(3)

Key Documents

367443

Sigma-Aldrich

2-Benzyloxy-1,3-propanediol

99%

Synonym(s):

2-O-Benzylglycerol, Glycerol 2-benzyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€181.00

€181.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€181.00

About This Item

Linear Formula:
C6H5CH2OCH(CH2OH)2
CAS Number:
Molecular Weight:
182.22
Beilstein:
1953926
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€181.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

bp

185-187 °C/10 mmHg (lit.)

mp

38-40 °C (lit.)

SMILES string

OCC(CO)OCc1ccccc1

InChI

1S/C10H14O3/c11-6-10(7-12)13-8-9-4-2-1-3-5-9/h1-5,10-12H,6-8H2

InChI key

UDIPIOHLDFSMLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service