Skip to Content
Merck
All Photos(1)

Key Documents

367168

Sigma-Aldrich

Tin(II) sulfide

96%

Synonym(s):

Herzenbergite, Stannous sulfide, Tin monosulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
SSn
CAS Number:
Molecular Weight:
150.78
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Assay:
96%

Assay

96%

density

5.22 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

S=[SnH2]

InChI

1S/S.Sn

InChI key

AFNRRBXCCXDRPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Cai Zhang et al.
Environmental science & technology, 45(21), 9324-9331 (2011-10-06)
SnS(2) nanocrystals with adjustable sizes were synthesized via a hydrothermal method from the aqueous solution of common and inexpensive SnCl(4)·5H(2)O, thioacetamide and citric acid, simply by varying the reaction temperature and reaction time. The structures, Brunauer-Emmett-Teller (BET) specific surface areas
Jiajia Ning et al.
Nanoscale, 2(9), 1699-1703 (2010-09-08)
SnS nanocrystals have been synthesized in a simple and facile way. Sn(6)O(4)(OH)(4) is introduced to synthesize tin sulfide, which is used as tin precursor. By changing the reaction conditions (reaction temperature and Sn/S molar ratio), SnS nanocrystals with different shape
Yang Zhang et al.
Nanotechnology, 24(2), 025503-025503 (2012-12-15)
We demonstrate a practical sensing platform, consisting of SnO(2) nanoparticle-decorated semiconducting single-walled carbon nanotubes assembled on gold electrodes via a dielectrophoretic process, for highly sensitive CO detection with fast response at room temperature. The highest sensitivity obtained was 0.27 and
Hiroki Tsukigase et al.
Journal of nanoscience and nanotechnology, 11(4), 3215-3221 (2011-07-23)
SnS2 nanoparticles were synthesized through a simple wet chemical process at room temperature. The SnS2 nanoparticles were approximately spherical in shape and had diameter about 3-4 nm. SnS2-sensitized TiO2 electrodes were fabricated by the immersion of chemically modified TiO2 to
Guozhang Dai et al.
Journal of the American Chemical Society, 132(35), 12174-12175 (2010-08-19)
Semiconductor superlattice micro-/nanowires could greatly increase the versatility and power of modulating electronic (or excitonic, photonic) transport, optical properties. In this communication, we report growth of a semiconductor CdS/CdS:SnS(2) superlattice microwire through a coevaporation technique with microenvironmental control. Such a

Questions

  1. What is the band gap of SnS

    1 answer
    1. The band gap of SnS is not determined. See the links below to review publications that suggest band gaps for bulk and film SnS, respectively.
      https://www.nature.com/articles/s41467-018-03897-3
      https://pubs.acs.org/doi/10.1021/jacs.5b08236

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service