Skip to Content
Merck
All Photos(2)

Key Documents

366188

Sigma-Aldrich

N-Benzylcinchoninium chloride

98%

Synonym(s):

9-Hydroxy-1-(phenylmethyl)cinchonanium chloride, BCNC

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€262.00

€262.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€262.00

About This Item

Empirical Formula (Hill Notation):
C26H29ClN2O
CAS Number:
Molecular Weight:
420.97
Beilstein:
5232950
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€262.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

optical activity

[α]20/D +169°, c = 0.4 in H2O

mp

256 °C (dec.) (lit.)

functional group

hydroxyl
phenyl

SMILES string

[Cl-].O[C@H]([C@H]1C[C@@H]2CC[N+]1(C[C@@H]2C=C)Cc3ccccc3)c4ccnc5ccccc45

InChI

1S/C26H29N2O.ClH/c1-2-20-18-28(17-19-8-4-3-5-9-19)15-13-21(20)16-25(28)26(29)23-12-14-27-24-11-7-6-10-22(23)24;/h2-12,14,20-21,25-26,29H,1,13,15-18H2;1H/q+1;/p-1/t20-,21-,25+,26-,28?;/m0./s1

InChI key

FCHYSBWCOKEPNQ-YGNISETGSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Phase-transfer catalyst involved in:
  • Enantioselective alkylation of malonic diester with alkyl halides
  • Hydrolysis of enol esters
  • Asymmetric 6π electrocyclization for synthesis of functionalized indolines
  • Asymmetric Michael addition
  • Alkylation of monosubstituted indolinones with alkylhalides
  • Asymmetric destruction and kinetic resolution of 1-benzylated Reissert compounds

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service