Skip to Content
Merck
All Photos(1)

Key Documents

366145

Sigma-Aldrich

2,3-Dichloro-5-(trifluoromethyl)pyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2Cl2F3N
CAS Number:
Molecular Weight:
215.99
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.475 (lit.)

bp

80 °C/20 mmHg (lit.)

density

1.549 g/mL at 25 °C (lit.)

functional group

chloro
fluoro

SMILES string

FC(F)(F)c1cnc(Cl)c(Cl)c1

InChI

1S/C6H2Cl2F3N/c7-4-1-3(6(9,10)11)2-12-5(4)8/h1-2H

InChI key

ABNQGNFVSFKJGI-UHFFFAOYSA-N

General description

2,3-Dichloro-5-(trifluoromethyl)pyridine is reported as an intermediate of herbicide. Synthesis of 2,3-dichloro-5-(trifluoromethyl)pyridine has been reported.[1] Palladium-catalyzed monoalkoxycarbonylation of 2,3-dichloro-5-(trifluoromethyl)pyridine has been reported.[2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic process development and scale up of palladium-catalyzed alkoxycarbonylation of chloropyridines.
Crettaz R, et al.
Organic Process Research & Development, 5(6), 572-574 (2001)
Research development of 2, 3-dichloro-5-trifluoromethyl-pyridine [J].
ZHAO W-K, et al.
Chemical Reagents, 6, 005-005 (2004)
Yong Xie et al.
Pest management science, 74(1), 189-199 (2017-08-02)
Agrochemicals have been crucial to the production of food, and the need for the development of novel agrochemicals continues unceasing owing to the loss of existing produces via the growth of resistance and the desire for products with more propitious

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service