Skip to Content
Merck
All Photos(2)

Key Documents

362964

Sigma-Aldrich

Calcium 2-ethylhexanoate

98%

Synonym(s):

2-Ethylhexanoic acid calcium salt, Calcium bis(2-ethyl hexanoate)

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€200.00

€200.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€200.00

About This Item

Linear Formula:
[CH3(CH2)3CH(C2H5)CO2]2Ca
CAS Number:
Molecular Weight:
326.49
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€200.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

lumps and powder

reaction suitability

core: calcium

SMILES string

CCCCC(CC)C(=O)O[Ca]OC(=O)C(CC)CCCC

InChI

1S/2C8H16O2.Ca/c2*1-3-5-6-7(4-2)8(9)10;/h2*7H,3-6H2,1-2H3,(H,9,10);/q;;+2/p-2

InChI key

LTPCXXMGKDQPAO-UHFFFAOYSA-L

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service