Skip to Content
Merck
All Photos(1)

Key Documents

361135

Sigma-Aldrich

2,5-Dimethylstyrene

contains 500 ppm tert-Butylcatechol as stabilizer, 98%

Synonym(s):

1,4-Dimethyl-2-ethenylbenzene, 1,4-Dimethyl-2-vinylbenzene, 2-Ethenyl-1,4-dimethylbenzene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€142.00

€142.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€142.00

About This Item

Linear Formula:
(CH3)2C6H3CH=CH2
CAS Number:
Molecular Weight:
132.20
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€142.00


Estimated to ship on29 May 2025


Request a Bulk Order

Assay

98%

form

liquid

contains

500 ppm tert-Butylcatechol as stabilizer

refractive index

n20/D 1.539 (lit.)

bp

71-72 °C/10 mmHg (lit.)

mp

−35 °C (lit.)

density

0.904 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1ccc(C)c(C=C)c1

InChI

1S/C10H12/c1-4-10-7-8(2)5-6-9(10)3/h4-7H,1H2,2-3H3

InChI key

DBWWINQJTZYDFK-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service