Skip to Content
Merck
All Photos(2)

Documents

360953

Sigma-Aldrich

α,α-Dicyanoethyl acetate

98%

Synonym(s):

α-Acetoxy-α-methylmalononitrile, DAC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C(CN)2CH3
CAS Number:
Molecular Weight:
138.12
Beilstein:
1763912
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

69-71 °C (lit.)

SMILES string

CC(=O)OC(C)(C#N)C#N

InChI

1S/C6H6N2O2/c1-5(9)10-6(2,3-7)4-8/h1-2H3

InChI key

OSNAAKXQIFQLNS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

α,α-Dicyanoethyl acetate may be used as a selective N-acetylation agent.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao Huang et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 84, 447-453 (2016-09-30)
Decitabine (5-aza-2'-deoxycytidine, DAC) and 5-azacitidine (Aza), an inhibitor of DNA methyltransferases, possess a wide range of anti-metabolic and anti-cancer activities. This study examined the effects of DAC and Aza on inflammatory and oxidative injuries, as well as on glycocalyx and
Simone Anzellini et al.
Scientific reports, 9(1), 13034-13034 (2019-09-12)
In this work, the melting line of platinum has been characterized both experimentally, using synchrotron X-ray diffraction in laser-heated diamond-anvil cells, and theoretically, using ab initio simulations. In the investigated pressure and temperature range (pressure between 10 GPa and 110 GPa and
α,α-Dicyanoethyl Acetate. A Reagent for Selective N-Acetylation.
Ho, T-L.
Synthetic Communications, 4(6), 351-353 (1974)
D Herbreteau et al.
AJNR. American journal of neuroradiology, 37(12), 2280-2286 (2016-08-20)
Endovascular treatment with the flow-disrupter Woven EndoBridge aneurysm embolization system (WEB) is an innovative treatment for wide-neck bifurcation aneurysms. Prospective, multicenter studies have shown the high safety of this technique. Stability of aneurysm occlusion in long-term follow-up has been rarely
Erol Akgul et al.
World neurosurgery, 95, 229-240 (2016-08-16)
The DERIVO embolization device (DED) is a new nitinol flow diverter stent manufactured for the treatment of intracranial aneurysms. In this study, we evaluated the safety and efficacy of the DED in the treatment of intracranial aneurysms and present the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service