Skip to Content
Merck
All Photos(2)

Key Documents

359599

Sigma-Aldrich

cis-4-Benzyloxy-2-buten-1-ol

≥95%

Synonym(s):

cis-2-Buten-1,4-diol-monobenzyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€171.00

€171.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
€171.00

About This Item

Linear Formula:
C6H5CH2OCH2CH=CHCH2OH
CAS Number:
Molecular Weight:
178.23
Beilstein:
3605116
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€171.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

liquid

refractive index

n20/D 1.534 (lit.)

bp

105-108 °C/0.1 mmHg (lit.)

density

1.058 g/mL at 25 °C (lit.)

functional group

ether
hydroxyl
phenyl

storage temp.

2-8°C

SMILES string

OC\C=C/COCc1ccccc1

InChI

1S/C11H14O2/c12-8-4-5-9-13-10-11-6-2-1-3-7-11/h1-7,12H,8-10H2/b5-4-

InChI key

CGLJRLXTVXHOLX-PLNGDYQASA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

cis-4-Benzyloxy-2-buten-1-ol may be used in the synthesis of cis-4-benzyloxybuten-2-ylamine via Mitsunobu reaction.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Novel 3-Substituted Pyrrolo[2,3-b]quinoxalines via an Intramolecular Heck Reaction on an Aminoquinoxaline Scaffold.
Jie Jack Li
The Journal of organic chemistry, 64(22), 8425-8427 (2001-10-25)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service