Skip to Content
Merck
All Photos(1)

Key Documents

359394

Sigma-Aldrich

Sodium mercaptopyruvate

≥85%, technical grade

Synonym(s):

3-Mercapto-2-oxopropionic acid sodium salt, Mercaptopyruvic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2COCOONa
CAS Number:
Molecular Weight:
142.11
Beilstein:
4018548
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

≥85%

form

powder

impurities

<5 wt. % ethanol

mp

200 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

[Na+].[O-]C(=O)C(=O)CS

InChI

1S/C3H4O3S.Na/c4-2(1-7)3(5)6;/h7H,1H2,(H,5,6);/q;+1/p-1

InChI key

CODUSAVZTZYYDD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium mercaptopyruvate is suitable for use in the thiosulfate sulfurtransferase activity assay.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Herbert T Nagasawa et al.
Journal of medicinal chemistry, 50(26), 6462-6464 (2007-11-28)
A series of prodrugs of 3-mercaptopyruvate (3-MP), the substrate for the enzyme 3-mercaptopyruvate/cyanide sulfurtransferase (3-MPST) that converts cyanide to the nontoxic thiocyanate, which are highly effective cyanide antidotes, have been developed. These prodrugs of 3-MP are unique in being not
J Papenbrock et al.
European journal of biochemistry, 267(1), 145-154 (1999-12-22)
A database search for similarities between sequenced parts of the Arabidopsis thaliana genome with known sulfurtransferase sequences from Escherichia coli and mammals was undertaken to obtain information about plant sulfurtransferase-like proteins. One gene and several homologous EST clones were identified.
T Nakamura et al.
European journal of biochemistry, 267(17), 5621-5630 (2000-08-22)
Mercaptopyruvate sulfurtransferase (MST, EC 2.8.1.2) and thiosulfate sulfurtransferase (TST, rhodanese, EC 2.8.1.1) are evolutionarily related enzymes that catalyze the transfer of sulfur ions from mercaptopyruvate and thiosulfate, respectively, to cyanide ions. We have isolated and characterized two cDNAs, AtMST1 and
N E Labrou et al.
Journal of chromatography. A, 718(1), 35-44 (1995-12-01)
Three biomimetic dye ligands bearing as a triazine-linked terminal moiety a carboxylated structure, which mimics substrates and inhibitors of L-lactate dehydrogenase (LDH), were immobilized on cross-linked agarose Ultrogel A6R. These biomimetic dyes are purpose-designed analogues of commercial monochlorotriazine Cibacron Blue
Steven E Patterson et al.
Journal of medicinal chemistry, 56(3), 1346-1349 (2013-01-11)
Current cyanide antidotes are administered by IV infusion, which is suboptimal for mass casualties. Therefore, in a cyanide disaster, intramuscular (IM) injectable antidotes would be more appropriate. We report the discovery of the highly water-soluble sulfanegen triethanolamine as a promising

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service