Skip to Content
Merck
All Photos(1)

Key Documents

358932

Supelco

3,3′-Dimethylnaphthidine

99%

Synonym(s):

4,4′-Diamino-3,3′-dimethyl-1,1′-binaphthyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20N2
CAS Number:
Molecular Weight:
312.41
Beilstein:
2152306
EC Number:
MDL number:
UNSPSC Code:
12161500
PubChem Substance ID:

Assay

99%

technique(s)

titration: suitable

SMILES string

Cc1cc(-c2cc(C)c(N)c3ccccc23)c4ccccc4c1N

InChI

1S/C22H20N2/c1-13-11-19(15-7-3-5-9-17(15)21(13)23)20-12-14(2)22(24)18-10-6-4-8-16(18)20/h3-12H,23-24H2,1-2H3

InChI key

FTEGMIZQLGXBNA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service