Skip to Content
Merck
All Photos(1)

Key Documents

357715

Sigma-Aldrich

2-Ethylpyrrole

technical grade, 90%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€72.30

€72.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€72.30

About This Item

Empirical Formula (Hill Notation):
C6H9N
CAS Number:
Molecular Weight:
95.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€72.30


Check Cart for Availability

Request a Bulk Order

grade

technical grade

Quality Level

Assay

90%

form

liquid

refractive index

n20/D 1.497 (lit.)

bp

164-165 °C/754 mmHg (lit.)

density

0.929 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCc1ccc[nH]1

InChI

1S/C6H9N/c1-2-6-4-3-5-7-6/h3-5,7H,2H2,1H3

InChI key

XRPDDDRNQJNHLQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Ethylpyrrole is a monosubstituted alkyl pyrrole. Stochastic, Rothemund-type condensation of pyrrole, 2-ethylpyrrole and arylaldehyde yields 3-ethyl-substituted inverted porphyrin, which readily converts to 3-(1′-hydroxyethyl)- and 3-acetyl-inverted porphyrins.[1] 2-Ethylpyrrole is a conjugated five-membered heterocyclic flavor compound and its photooxidation reactivity was investigated.[2]

Application

2-Ethylpyrrole may be used in the preparation of 1-(3-methoxyphenyl)-2-ethylpyrrole.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Application of 2-ethylpyrrole for a direct synthesis of 3-substituted inverted porphyrins.
Schmidt I and Chmielewski PJ.
Tetrahedron Letters, 42(36), 6389-6392 (2001)
Reactivity on photooxidation of selected five-membered heterocyclic flavor compounds.
Chen C-W and Ho C-T.
Journal of Agricultural and Food Chemistry, 44(8), 2078-2080 (1996)
Jon C Antilla et al.
The Journal of organic chemistry, 69(17), 5578-5587 (2004-08-17)
This paper details the copper-catalyzed N-arylation of pi-excessive nitrogen heterocycles. The coupling of either aryl iodides or aryl bromides with common nitrogen heterocycles (pyrroles, pyrazoles, indazoles, imidazoles, and triazoles) was successfully performed in good yield with catalysts derived from diamine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service