Skip to Content
Merck
All Photos(2)

Key Documents

357472

Sigma-Aldrich

Sodium aluminum hydride

technical grade, 90%

Synonym(s):

Sodium tetrahydroaluminate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaAlH4
CAS Number:
Molecular Weight:
54.00
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

reaction suitability

reagent type: reductant

SMILES string

[Na+].[AlH4-]

InChI

1S/Al.Na.4H/q-1;+1;;;;

InChI key

MNYCIYRTKITUKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant used for hydrogen storage
Sodium aluminum hydride (NaAlH4) can be used as a hydrogen storage material with a hydrogen content of 5.6%. It can be doped with titanium (Ti) for further use in reversible hydrogen storage.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B - Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Ismail, et al.,
International Journal of Hydrogen Energy, 36, 9045-9050 (2011)
T. Ono, et al.,
J. Alloy Compounds, 506, 297-301 (2010)
Gugelchuk M
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2009)
Effect of graphite as a co-dopant on the dehydrogenation and hydrogenation kinetics of Ti-doped sodium aluminum hydride
Wang J, et al.
Journal of alloys and compounds, 395(1-2), 252-262 (2005)
Carbon nanostructures as catalyst for improving the hydrogen storage behavior of sodium aluminum hydride
Hudson MSL, et al.
International Journal of Hydrogen Energy, 37(3), 2750-2755 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service