Skip to Content
Merck
All Photos(1)

Key Documents

349712

Sigma-Aldrich

Benzyl chloroacetate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CO2CH2C6H5
CAS Number:
Molecular Weight:
184.62
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

refractive index

n20/D 1.525 (lit.)

bp

90 °C/1 mmHg (lit.)

density

1.215 g/mL at 25 °C (lit.)

functional group

chloro
ester
phenyl

SMILES string

ClCC(=O)OCc1ccccc1

InChI

1S/C9H9ClO2/c10-6-9(11)12-7-8-4-2-1-3-5-8/h1-5H,6-7H2

InChI key

SOGXBRHOWDEKQB-UHFFFAOYSA-N

General description

Benzyl chloroacetate is an ester.

Application

Benzyl chloroacetate may be used:
  • in the synthesis of monoesters of phosphonoacetic acid
  • in the synthesis of N-benzoyl-glycyl- hydroxyl acetic acid, ester substrate for carboxypeptidase Y catalyzed peptide synthesis
  • as reagent in the total syntheses of the (±)-di-O-methyl ethers of the norlignans sequirin-A, agatharesinol, and hinokiresinol, and of (±)-tri-O-methyl sequirin-E

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective total syntheses of the (?)-di-O-methyl ethers of agatharesinol, sesquirin-A, and hinokiresinol, and of (?)-tri-O-methylsequirin-E, characteristic norlignans of coniferae.
Beracierta AP and Whiting DA.
Journal of the Chemical Society. Perkin Transactions 1, 10, 1257-1263 (1978)
Influence of the substrate structure on carboxypeptidase Y catalyzed peptide bond formation.
Breddam K, et al.
Carlsberg Research Communications, 45(5), 361-367 (1980)
T R Herrin et al.
Journal of medicinal chemistry, 20(5), 660-663 (1977-05-01)
The synthesis of monoesters (P and C) of phosphonoacetic acid (PA) is given. The carboxyl esters were prepared by two methods: the reaction of chloroacetates with tris(trimethylsilyl) phosphite, followed by hydrolysis; and by the acid-catalyzed esterification of PA with the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service