Skip to Content
Merck
All Photos(1)

Key Documents

347744

Sigma-Aldrich

5-Phenyl-1H-tetrazole

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6N4
CAS Number:
Molecular Weight:
146.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

mp

216 °C (dec.) (lit.)

functional group

phenyl

SMILES string

c1ccc(cc1)-c2nnn[nH]2

InChI

1S/C7H6N4/c1-2-4-6(5-3-1)7-8-10-11-9-7/h1-5H,(H,8,9,10,11)

InChI key

MARUHZGHZWCEQU-UHFFFAOYSA-N

General description

Nucleophilic substitution reactions on 1,4-dimethoxybenzene and 1,3,5-trimethoxybenzene by the anions of 5-phenyl-1H-tetrazole has been investigated by paired electrosynthesis.[1]

Application

5-Phenyl-1H-tetrazole may be used as ligand in the solvothermal synthesis of 3D polyoxometalate-based silver coordination polymers.[2] It was used in the synthesis of 5-phenyltetrazolate organolanthanide complexes.[3] It was used to investigate the corrosion inhibition of copper in salt water.[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Corrosion inhibition of copper in non-polluted and polluted sea water using 5-phenyl-1-H-tetrazole.
Al Kharafi FM, et al.
International Journal of Electrochemical Science, 7, 3289-3298 (2012)
Min-Xia Liang et al.
Inorganic chemistry, 53(2), 897-902 (2014-01-08)
Four 3D POM-based silver coordination polymers, namely, [Ag17(ptz)11(PW12O40)2]n (1), [Ag17(ptz)11(PMo12O40)2]n (2), [Ag12(ptz)6(CN)2(SiW12O40)]n (3), and [Ag19(ptz)8(H2ptz)(H3ptz)(AgP5W30O110)·7H2O]n (4), have been obtained by solvothermal reaction of AgNO3 and 5-phenyl-1H-tetrazole (Hptz) ligand in the presence of four types of polyoxometalates. Structural analysis shows that
A novel bonding mode of tetrazolate ligand to a metal: synthesis and structural characterization of 5-phenyltetrazolate organolanthanide complexes:
Zhou X-G, et al.
Journal of Organometallic Chemistry, 563(1), 101-112 (1998)
Nucleophilic aromatic substitution by paired electrosynthesis: Reactions of methoxy arenes with 1H-tetrazoles.
Hu K, et al.
Tetrahedron Letters, 36(39), 7027-7030 (1995)
David Furkert et al.
Cell chemical biology, 27(8), 1097-1108 (2020-08-14)
The inositol pyrophosphates (PP-InsPs) are a ubiquitous group of highly phosphorylated eukaryotic messengers. They have been linked to a panoply of central cellular processes, but a detailed understanding of the discrete signaling events is lacking in most cases. To create

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service