Skip to Content
Merck
All Photos(1)

Key Documents

347485

Sigma-Aldrich

2-(Trimethylsilyloxy)ethyl methacrylate

contains ≤100 ppm BHT as inhibitor, 96%

Synonym(s):

2-(Trimethoxysilyl)ethyl methacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CO2CH2CH2OSi(CH3)3
CAS Number:
Molecular Weight:
202.32
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

vapor pressure

<5 mmHg ( 25 °C)

Quality Level

Assay

96%

form

liquid

contains

≤100 ppm BHT as inhibitor

refractive index

n20/D 1.428 (lit.)

density

0.928 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(=O)OCCO[Si](C)(C)C

InChI

1S/C9H18O3Si/c1-8(2)9(10)11-6-7-12-13(3,4)5/h1,6-7H2,2-5H3

InChI key

WUGOQZFPNUYUOO-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Monika Zygo et al.
Nanomaterials (Basel, Switzerland), 10(3) (2020-03-28)
This study reports the utilization of controlled radical polymerization as a tool for controlling the stimuli-responsive capabilities of graphene oxide (GO) based hybrid systems. Various polymer brushes with controlled molecular weight and narrow molecular weight distribution were grafted from the
Rizacan Sarikaya et al.
Polymers, 12(9) (2020-09-17)
The inherent degradation property of most dental resins in the mouth leads to the long-term release of degradation by-products at the adhesive/tooth interface. The by-products increase the virulence of cariogenic bacteria, provoking a degradative positive-feedback loop that leads to physicochemical
D S Spencer et al.
International journal of pharmaceutics, 588, 119691-119691 (2020-07-30)
Cationic polymers have garnered significant interest for their utility in intracellular drug delivery and gene therapy. However, due to their associated toxicities, novel synthesis approaches must be explored to develop materials that are biocompatible. The novel library of nanoparticles synthesized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service