Skip to Content
Merck
All Photos(1)

Key Documents

34683

Sigma-Aldrich

4′,4″(5″)-Di-tert-butyldicyclohexano-18-crown-6

technical, mixture of isomers, ≥90.0% (GC)

Synonym(s):

2,13(or 2,14)-Bis(1,1-dimethylethyl)eicosahydrodibenzo[b,k][1,4,7,10,13,16]hexaoxacyclooctadecin, 4,4′(5′)-Bis(tert-butylcyclohexano)-18-crown-6, 4,4′(5′)-Di-t-butylcyclohexano-18-crown-6

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€399.00

€399.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€399.00

About This Item

Empirical Formula (Hill Notation):
C28H52O6
CAS Number:
Molecular Weight:
484.71
Beilstein:
3626927
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€399.00


Check Cart for Availability

Request a Bulk Order

Quality Level

grade

technical

Assay

≥90.0% (GC)

refractive index

n20/D 1.485

bp

190-195 °C/0.05 mmHg (lit.)

SMILES string

CC(C)(C)C1CCC2OCCOCCOC3CCC(CC3OCCOCCOC2C1)C(C)(C)C

InChI

1S/C28H52O6/c1-27(2,3)21-7-9-23-25(19-21)33-17-13-30-14-18-34-26-20-22(28(4,5)6)8-10-24(26)32-16-12-29-11-15-31-23/h21-26H,7-20H2,1-6H3

InChI key

SHJRQDXUSZYABF-UHFFFAOYSA-N

Application

  • Conveniently synthesis of porous crown-based resin with efficient 90Sr capture from highly acidic wastewater: This study discusses the synthesis of a porous crown-based resin incorporating 4′,4″(5″)-Di-tert-butyldicyclohexano-18-crown-6, which demonstrates efficient capture of 90Sr from highly acidic wastewater. The article details the performance and applications of the resin in nuclear waste management and environmental protection (Y Su, J He, X Liu, S Jia, K Hu, J Yang, T Liu… - Separation and Purification Technology, 2024).

Other Notes

Lipophilic crown ether; selective extraction of strontium[1]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Separation and preconcentration of strontium from biological, environmental, and nuclear waste samples by extraction chromatography using a crown ether.
E P Horwitz et al.
Analytical chemistry, 63(5), 522-525 (1991-03-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service