Skip to Content
Merck
All Photos(1)

Key Documents

346578

Sigma-Aldrich

5-Phenyl-1,3-cyclohexanedione

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H7(=O)2
CAS Number:
Molecular Weight:
188.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

mp

188 °C (lit.)

functional group

ketone
phenyl

SMILES string

O=C1CC(CC(=O)C1)c2ccccc2

InChI

1S/C12H12O2/c13-11-6-10(7-12(14)8-11)9-4-2-1-3-5-9/h1-5,10H,6-8H2

InChI key

UPPYKNLSSLIIAZ-UHFFFAOYSA-N

General description

5-Phenyl-1,3-cyclohexanedione is a 1,3-diketone.[1] Reaction of 5-phenyl-1,3-cyclohexanedione with N-substituted isatins in pyrindine has been investigated.[2]

Application

5-Phenyl-1,3-cyclohexanedione may be used:
  • in the synthesis of hexahydrobenzo[a]phenanthridin-4-one derivatives via condensation with N-arylmethylene-2-naphthylamines[3]
  • in the preparation of benzophenanthridine derivatives[1]
  • in the preparation of iodonium betaine[4]
  • in the synthesis of various 2H-pyrans by iodine-catalyzed reactions[5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chunhui Dai et al.
Beilstein journal of organic chemistry, 8, 986-993 (2012-09-29)
A mild and practical synthesis of spirooxindole [1,3]oxazino derivatives from N-substituted isatins and 1,3-dicarbonyl compounds with pyridine derivatives is reported. The reactions provided good to excellent yields. Further exploration of the molecular diversity of these compounds is demonstrated through Diels-Alder
Reactions of bisazomethines of the naphtalene series with 1, 3-diketones.
Kozlov SN, et al.
Russ. J. Gen. Chem., 73(9), 1424-1440 (2009)
Iodine-Catalyzed One-Pot Synthesis of 2H-Pyrans by Domino Knoevenagel/6p-Electrocylization.
Jung EJ, et al.
Bull. Korean Chem. Soc., 30, 2833-2836 (2009)
Efforts toward the synthesis of liphatic iodonium salts.
Dence JB and Roberts JD.
The Journal of Organic Chemistry, 33(3), 1251-1253 (1968)
Synthesis of Benzo [a] phenanthridine Derivatives by Condensation of N-Arylmethylene-2-naphthylamines with 5-Phenyl-and 5-(p-Methoxyphenyl)-1, 3-cyclohexanediones.
Kozlov NG, et al.
Russ. J. Gen. Chem., 72(8), 1238-1242 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service