Skip to Content
Merck
All Photos(2)

Key Documents

345059

Sigma-Aldrich

4-Fluoro-2-nitrophenol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(NO2)OH
CAS Number:
Molecular Weight:
157.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

75-77 °C (lit.)

functional group

fluoro
nitro

SMILES string

Oc1ccc(F)cc1[N+]([O-])=O

InChI

1S/C6H4FNO3/c7-4-1-2-6(9)5(3-4)8(10)11/h1-3,9H

InChI key

ZHRLVDHMIJDWSS-UHFFFAOYSA-N

General description

4-Fluoro-2-nitrophenol is formed during iron(III) nitrate nonahydrate activated by tungstophosphoric acid cesium salt catalyzed nitration of 4-fluorophenol.[1]

Application

4-Fluoro-2-nitrophenol may be used in the preparation of 5-fluoro-2-methoxyaniline[2]and 5-fluoro-2-hydroxyaniline.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalytic effects of some Keggin-type heteropoly acids and polyoxometalates on selective nitration of phenols.
Amani K and Maleki F.
Journal of the Iranian Chemical Society, 4(2), 238-243 (2007)
I M Rietjens et al.
European journal of biochemistry, 194(3), 945-954 (1990-12-27)
Pathways for biodehalogenation of fluorinated aniline derivatives were investigated. Microsomal NADPH-dependent dehalogenation of fluoroanilines was shown to proceed by three different reaction pathways. The first route appeared to result in monooxygenation at a fluorinated position and release of the fluorine
SOME SYNTHESES FROM p-FLUOROANISOLE.
BUU-HOI NP, et al.
The Journal of Organic Chemistry, 19(10), 1617-1621 (1954)
J I Cacho et al.
Journal of chromatography. A, 1456, 27-33 (2016-06-19)
A rapid and sensitive procedure for the determination of six NPs in soils by gas chromatography and mass spectrometry (GC-MS) is proposed. Ultrasound assisted extraction (UAE) is used for NP extraction from soil matrices to an organic solvent, while the
Marta Pastor-Belda et al.
Talanta, 189, 543-549 (2018-08-09)
This paper presents a procedure for the determination of seven nitrophenols (NPs) in water and soil samples using stir bar sorptive extraction (SBSE) coupled to gas chromatography with mass spectrometry (GC-MS) by means of a thermal desorption unit (TDU). Microwave

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service