Skip to Content
Merck
All Photos(3)

Key Documents

344370

Sigma-Aldrich

Lithium methoxide

98%

Synonym(s):

Lithium methylate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€103.00
100 G
€313.00

€103.00


Estimated to ship on09 April 2025


Request a Bulk Order

Select a Size

Change View
25 G
€103.00
100 G
€313.00

About This Item

Linear Formula:
CH3OLi
CAS Number:
Molecular Weight:
37.97
Beilstein:
3552389
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€103.00


Estimated to ship on09 April 2025


Request a Bulk Order

Quality Level

Assay

98%

form

powder

reaction suitability

core: lithium

SMILES string

[Li+].C[O-]

InChI

1S/CH3O.Li/c1-2;/h1H3;/q-1;+1

InChI key

JILPJDVXYVTZDQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Employed in sol-gel synthesis of lithium borophosphates.[1]

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-heat. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

68.0 °F

Flash Point(C)

20 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali, A.F. et al.
Journal of Materials Research, 14, 1510-1510 (1999)

Articles

Few Monolayer Atomic Layer Deposition (ALD) on Surfaces and Interfaces for Energy Applications

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service