Skip to Content
Merck
All Photos(1)

Key Documents

342262

Sigma-Aldrich

Potassium hexachloroiridate(III)

Synonym(s):

Tripotassium hexachloroiridate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€183.00

€183.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€183.00

About This Item

Linear Formula:
K3IrCl6
CAS Number:
Molecular Weight:
522.23
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
solid

€183.00


Check Cart for Availability

Request a Bulk Order

Assay

35.5-38.1% iridium (Ir) basis (after reduction with magnesium, gravimetric)

Quality Level

form

solid

concentration

35.5-38.1% Ir (after reduction with magnesium, gravimetric)

SMILES string

[K+].[K+].[K+].Cl[Ir-3](Cl)(Cl)(Cl)(Cl)Cl

InChI

1S/6ClH.Ir.3K/h6*1H;;;;/q;;;;;;+3;3*+1/p-6

InChI key

NZKWZUOYGAKOQC-UHFFFAOYSA-H

Related Categories

Application

  • Stability Enhancement of Galvanic Redox Potentiometry by Optimizing the Redox Couple in Counterpart Poles: Discusses the enhancement of redox potentiometry stability using potassium hexachloroiridate (Wei et al., 2023).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iva Turyan et al.
Analytical biochemistry, 494, 108-113 (2015-11-10)
Measuring and monitoring of protein oxidation modifications is important for biopharmaceutical process development and stability assessment during long-term storage. Currently available methods for biomolecules oxidation analysis use time-consuming peptide mapping analysis. Therefore, it is desirable to develop high-throughput methods for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service