Skip to Content
Merck
All Photos(2)

Key Documents

34072

Sigma-Aldrich

2,4-Dibromobutyryl chloride

technical, ~90% (GC)

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
€221.00
50 ML
€772.00

€221.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
€221.00
50 ML
€772.00

About This Item

Linear Formula:
BrCH2CH2CHBrCOCl
CAS Number:
Molecular Weight:
264.34
Beilstein:
4955401
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€221.00


Check Cart for Availability

Request a Bulk Order

grade

technical

Assay

~90% (GC)

refractive index

n20/D 1.535

density

2.00 g/mL at 20 °C

functional group

acyl chloride
bromo

storage temp.

2-8°C

SMILES string

ClC(=O)C(Br)CCBr

InChI

1S/C4H5Br2ClO/c5-2-1-3(6)4(7)8/h3H,1-2H2

InChI key

WYZLYWUZERABRL-UHFFFAOYSA-N

Application

2,4-Dibromobutyryl chloride was used in the preparation of:
  • methyl 2,4-dibromobutanoate[1]
  • series of 3-(3, 5-di-tert-butyl-4-hydroxybenzylidene)pyrrolidin-2-ones, anti-inflammatory/analgesic agents[2]
  • α-bromolactam[3]

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of enantiopure 2-acylazetidines and their reactions with chloroformates.
Ma S, et al.
Tetrahedron Letters, 48(2), 269-271 (2007)
H Ikuta et al.
Journal of medicinal chemistry, 30(11), 1995-1998 (1987-11-01)
A series of 3-(3,5-di-tert-butyl-4-hydroxybenzylidene)pyrrolidin-2-ones was synthesized and evaluated as candidate antiinflammatory/analgesic agents as well as dual inhibitors of prostaglandin and leukotriene synthesis. Some compounds that showed dual inhibitory activity were found to possess equipotent antiinflammatory activities to indomethacin, with reduced
Bhooma Raghavan et al.
The Journal of organic chemistry, 71(5), 2151-2154 (2006-02-25)
A concise, stereoselective synthesis of alpha-substituted gamma-lactams is reported. gamma-Lactam scaffolds 2 and 3, possessing an Evans' chiral auxiliary and two types of N substituents, were successfully alkylated with different electrophiles to give alpha-substituted gamma-lactams with reasonable diastereoselectivities. The use

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service