340707
Manganese(II) nitrate solution
48-52 wt. % in dilute nitric acid
Synonym(s):
Manganese dinitrate, Manganous nitrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
liquid
composition
HNO3, <5 wt. %
concentration
45-50 wt. % (complexometric)
48-52 wt. % in dilute nitric acid
density
1.536 g/mL at 25 °C
SMILES string
[Mn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
InChI
1S/Mn.2NO3/c;2*2-1(3)4/q+2;2*-1
InChI key
MIVBAHRSNUNMPP-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 2 - Skin Corr. 1C - STOT RE 2 Inhalation
Target Organs
Brain
Supplementary Hazards
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Journal of hazardous materials, 357, 332-340 (2018-06-16)
To achieve efficient degradation of toluene, a series of Cu1-yMn2CeyOx/sepiolite catalysts (y = 0.1, 0.2, and 0.3) with different Cu1-yMn2CeyOx loadings (10%, 20%, and 30%) were prepared via the co-precipitation method. The structure-activity and surficial elemental species of Cu1-yMn2CeyOx/sepiolite were characterized by
Drug development and industrial pharmacy, 46(5), 846-851 (2020-04-18)
In The present project, a variety of MnFe2O4 (Mn) and Cr2Fe6O12 (Cr)-based nanocarriers (NCs) were synthesized as photosensitizer and NCs for delivery of chemotherapeutic curcumin (CUR) and provide a new structure for Photodynamic Therapy (PDT). For determining efficiency of NCs
ChemSusChem, 9(21), 3102-3112 (2016-10-13)
The synthesis of terephthalic acid from biomass remains an unsolved challenge. In this study, we conducted the selective oxidation of p-cymene (synthesized from biodegradable terpenes, limonene, or eucalyptol) into terephthalic acid over a Mn-Fe mixed-oxide heterogeneous catalyst. The impact of
The Science of the total environment, 703, 134781-134781 (2019-11-21)
As Prussian Blue analogues (PBAs) represent one of the most classical families of coordination compounds and exhibit versatile catalytic activities, PBAs have been considered as useful heterogeneous catalysts for reduction of 4-nitrophenol (4-NP) to 4-aminophenol (4-AP). Nevertheless, while Cu has
Environmental science and pollution research international, 26(10), 10117-10126 (2019-02-13)
Manganese (Mn)-modified ferric oxide/nickel foam (Fe/Ni) catalysts were prepared using Ni as a carrier, Fe and Mn as active components to study NH3-SCR of NOx at low temperature. The effects of different Fe loads and Mn-modified Fe/Ni catalysts on the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service