Skip to Content
Merck
All Photos(2)

Key Documents

340014

Sigma-Aldrich

Anthrarufin

technical grade, 85%

Synonym(s):

1,5-Dihydroxyanthraquinone

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€144.00

€144.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€144.00

About This Item

Empirical Formula (Hill Notation):
C14H8O4
CAS Number:
Molecular Weight:
240.21
Beilstein:
1881718
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€144.00


Check Cart for Availability

Request a Bulk Order

grade

technical grade

vapor density

8.3 (vs air)

Assay

85%

form

solid

mp

279 °C (dec.) (lit.)

SMILES string

Oc1cccc2C(=O)c3c(O)cccc3C(=O)c12

InChI

1S/C14H8O4/c15-9-5-1-3-7-11(9)14(18)8-4-2-6-10(16)12(8)13(7)17/h1-6,15-16H

InChI key

JPICKYUTICNNNJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katarzyna Lech et al.
Molecules (Basel, Switzerland), 25(14) (2020-07-19)
Effective analytical approaches for the identification of natural dyes in historical textiles are mainly based on high-performance liquid chromatography coupled with spectrophotometric detection and tandem mass spectrometric detection with electrospray ionization (HPLC-UV-Vis-ESI MS/MS). Due to the wide variety of dyes
Atsushi Furuta et al.
International journal of molecular sciences, 16(8), 18439-18453 (2015-08-12)
Hepatitis C virus (HCV) is an important etiological agent of severe liver diseases, including cirrhosis and hepatocellular carcinoma. The HCV genome encodes nonstructural protein 3 (NS3) helicase, which is a potential anti-HCV drug target because its enzymatic activity is essential

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service