Skip to Content
Merck
All Photos(1)

Documents

337919

Sigma-Aldrich

Piperonyl acetate

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O4
CAS Number:
Molecular Weight:
194.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥99%

refractive index

n20/D 1.5265 (lit.)

bp

150-151 °C/10 mmHg (lit.)

density

1.227 g/mL at 25 °C (lit.)

SMILES string

CC(=O)OCc1ccc2OCOc2c1

InChI

1S/C10H10O4/c1-7(11)12-5-8-2-3-9-10(4-8)14-6-13-9/h2-4H,5-6H2,1H3

InChI key

PFWYHTORQZAGCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Piperonyl acetate is a member of fragrance structural group aryl alkyl alcohol simple acid ester. It is a food additive.

Application

Piperonyl acetate was used as fragrance ingredient and its toxicologic and dermatologic review has been reported.

comparable product

Product No.
Description
Pricing

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D McGinty et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50 Suppl 2, S358-S362 (2012-03-27)
A toxicologic and dermatologic review of piperonyl acetate when used as a fragrance ingredient is presented. Piperonyl acetate is a member of the fragrance structural group aryl alkyl alcohol simple acid esters (AAASAE). The AAASAE fragrance ingredients are prepared by
D J Wright et al.
Xenobiotica; the fate of foreign compounds in biological systems, 10(4), 265-270 (1980-04-01)
1. The metabolism of piperonylidene acetone, and the food additives piperonyl acetone, piperonyl acetate and piperonyl isobutyrate were studied in male rabbits after oral doses of 100 mg/kg. 2. Both piperonylidene acetone and piperonyl acetone were extensively excreted as 4-(3,4-methylenedioxyphenyl)-butan-2-ol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service