Skip to Content
Merck
All Photos(2)

Key Documents

336734

Sigma-Aldrich

Ammonium tetrathiotungstate

≥99.9% trace metals basis

Synonym(s):

Ammonium tungsten sulfide ((NH4 )2 WS4 ), Bis(ammonium) tetrathiotungstate(2-) , Diammonium tetrathiotungstate, Diammonium tetrathiotungstate(2-)

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€202.00
25 G
€721.00

€202.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€202.00
25 G
€721.00

About This Item

Linear Formula:
(NH4)2WS4
CAS Number:
Molecular Weight:
348.18
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

€202.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99.9% trace metals basis

form

crystalline

density

2.71 g/mL at 25 °C (lit.)

SMILES string

N.N.S[W](S)(=S)=S

InChI

1S/2H3N.2H2S.2S.W/h2*1H3;2*1H2;;;/q;;;;;;+2/p-2

InChI key

MZSROLOWPFQNGM-UHFFFAOYSA-L

Related Categories

Application

This classic building block for transition metal sulfide cluster compounds was recently used in a low-temperature, solid state synthesis of a new series of heterometallic sulfide clusters stabilized with dppm [bis(diphenylphosphino)methane] ligands.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guoqing Hou et al.
Translational research : the journal of laboratory and clinical medicine, 149(5), 260-264 (2007-05-01)
Tetrathiomolybdate (TM) is an anticopper drug under development for treating Wilson's disease. Its mechanism of action involves forming a tight tripartite complex in the blood with serum albumin and available copper. When available copper levels are lowered in animals with
Shu Min Tan et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(35), 8510-8519 (2017-04-28)
A multitude of research into the application of transition-metal dichalcogenides as earth-abundant hydrogen evolution reaction (HER) electrocatalysts has been conducted. However, current synthetic methods generally deploy environmentally harmful chemicals and energy-consuming reaction conditions, despite the primary intent to attain renewable
Won-Hee Ryu et al.
ACS nano, 10(3), 3257-3266 (2016-01-26)
Heterogeneous electrode materials with hierarchical architectures promise to enable considerable improvement in future energy storage devices. In this study, we report on a tailored synthetic strategy used to create heterogeneous tungsten sulfide/oxide core-shell nanofiber materials with vertically and randomly aligned
B W Young et al.
Journal of inorganic biochemistry, 16(2), 121-134 (1982-04-01)
The effects of dietary supplementation with ammonium tetrathiotungstate and dithiotungstate on copper metabolism in young rats have been investigated. The addition of WS4(2-) (4-32 mg W/kg diet) decreased growth rates and induced clinical and biochemical signs of Cu deficiency. These
Kyung-Hee Park et al.
Chemical & pharmaceutical bulletin, 54(9), 1266-1270 (2006-09-02)
Tyrosinase requires two copper ions at the active site, in order to oxidize phenols to catechols. In this study, the inhibitory effect of the copper-chelating compound, ammonium tetrathiotungstate (ATTT), on the tyrosinase activity was investigated. ATTT was determined to inactivate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service