Skip to Content
Merck
All Photos(1)

Key Documents

336521

Sigma-Aldrich

4,4-Dimethyl-2-cyclopenten-1-one

99%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€581.00

€581.00


Estimated to ship on18 April 2025


Request a Bulk Order

Select a Size

Change View
1 G
€581.00

About This Item

Linear Formula:
(CH3)2C5H4(=O)
CAS Number:
Molecular Weight:
110.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€581.00


Estimated to ship on18 April 2025


Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.457 (lit.)

bp

158 °C (lit.)

density

0.903 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

CC1(C)CC(=O)C=C1

InChI

1S/C7H10O/c1-7(2)4-3-6(8)5-7/h3-4H,5H2,1-2H3

InChI key

YVFVCSCZJJGBAK-UHFFFAOYSA-N

Related Categories

General description

4,4-Dimethyl-2-cyclopenten-1-one is an α,β-unsaturated carbonyl compound and was reported to exhibit the higher tumor-specific cytotoxicity against oral human normal and tumor cells.[1] 4,4-Dimethyl-2-cyclopenten-1-one was identified as one of the constituents in the essential oil of Tagetes minuta L by GC-MS.[2]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chemical composition of the essential oil of Tagetes minuta growing in Saudi Arabia.
El-Deeb KS, et al.
Saudi Pharmaceutical Journal : SPJ : The Official Publication of the Saudi Pharmaceutical Society, 12(1), 51-53 (2004)
Tohru Nakayachi et al.
Anticancer research, 24(2B), 737-742 (2004-05-27)
A series of simple alpha, beta-unsaturated carbonyl compounds (1-26) was characterized for their cytotoxic profiles against oral human normal and tumor cells. Several cycloalkenones showed potent cytotoxic activities against human oral squamous cell carcinoma HSC-2 cell line. Among them, 4,4-dimethyl-2-cyclopenten-1-one

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service