Skip to Content
Merck
All Photos(1)

Key Documents

336351

Sigma-Aldrich

Zirconium(IV) carbide

powder, 10 μm, 99%

Synonym(s):

Zirconium monocarbide, Zirconium monocarbide (ZrC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZrC
CAS Number:
Molecular Weight:
103.23
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

description

cubic phase

Assay

99%

form

powder

particle size

10 μm

density

6.73 g/mL at 25 °C (lit.)

SMILES string

[C-]#[Zr+]

InChI

1S/C.Zr/q-1;+1

InChI key

MHMXYYDKIQTOHA-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service