Skip to Content
Merck
All Photos(1)

Key Documents

33624

Sigma-Aldrich

(−)-O,O′-Dibenzoyl-L-tartaric acid mono(dimethylamide)

≥99.0%

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€102.00

€102.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€102.00

About This Item

Empirical Formula (Hill Notation):
C20H19NO7
CAS Number:
Molecular Weight:
385.37
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

€102.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99.0%

optical activity

[α]20/D −79±1°, c = 5% in ethanol

mp

150-152 °C (dec.)

functional group

amide
carboxylic acid
ester
phenyl

SMILES string

CN(C)C(=O)[C@H](OC(=O)c1ccccc1)[C@@H](OC(=O)c2ccccc2)C(O)=O

InChI

1S/C20H19NO7/c1-21(2)17(22)15(27-19(25)13-9-5-3-6-10-13)16(18(23)24)28-20(26)14-11-7-4-8-12-14/h3-12,15-16H,1-2H3,(H,23,24)/t15-,16-/m1/s1

InChI key

LINPSOODYGSBAH-HZPDHXFCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chiral acid useful for the resolution of amines

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service