Skip to Content
Merck
All Photos(1)

Key Documents

334871

Sigma-Aldrich

3-Fluoro-p-anisidine

98%

Synonym(s):

3-Fluoro-4-methoxyaniline

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€77.80

€77.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€77.80

About This Item

Linear Formula:
FC6H3(OCH3)NH2
CAS Number:
Molecular Weight:
141.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€77.80


Check Cart for Availability

Request a Bulk Order

Assay

98%

bp

135 °C/18 mmHg (lit.)

mp

81-83 °C (lit.)

functional group

fluoro

SMILES string

COc1ccc(N)cc1F

InChI

1S/C7H8FNO/c1-10-7-3-2-5(9)4-6(7)8/h2-4H,9H2,1H3

InChI key

LJWAPDSCYTZUJU-UHFFFAOYSA-N

General description

Arrhenius parameter has been measured for the reactions of 3-fluoro-p-anisidine (3-fluoro-4-methoxyaniline) with picryl chloride.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The kinetics of the reactions of picryl chloride with some substituted anilines. Part 6. 4-Substituted and 3, 4-disubstituted anilines.
Emokpae TA, et al.
J. Chem. Soc. Perkin Trans. II, 6, 823-834 (1980)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service